Assay Details
Gene Symbol: LOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250756.1 | NP_001237685.1 | 6 - 7 | 1337 | 100 | |
GenBank mRNA | D13949.1 | - | 6 - 7 | 1337 | 100 | |
J03211.1 | - | 6 - 7 | 1363 | 100 | ||
JQ929619.1 | - | 6 - 7 | 1282 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
