Assay Details
Gene Symbol: GR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251077.1 | NP_001238006.1 | 4 - 5 | 1065 | 99 | |
GenBank mRNA | L11632.1 | - | 4 - 5 | 1065 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
