Assay Details
Gene Symbol: SRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251240.1 | NP_001238169.1 | 7 - 8 | 691 | 90 | |
GenBank mRNA | BT091931.1 | - | 7 - 8 | 630 | 90 | |
L27417.1 | - | 7 - 8 | 691 | 90 | ||
U58854.1 | - | 7 - 8 | 612 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
