Assay Details
Gene Symbol: TBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251273.1 | NP_001238202.1 | 6 - 7 | 503 | 98 | |
GenBank mRNA | L28002.1 | - | 6 - 7 | 503 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
