Assay Details
| Gene Symbol: RPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250039.1 | NP_001236968.1 | 5 - 6 | 443 | 100 | |
| GenBank mRNA | AK286733.1 | - | 5 - 6 | 466 | 100 | |
| BT089187.1 | - | 5 - 6 | 443 | 100 | ||
| M31024.1 | - | 3 - 4 | 333 | 100 | ||
More Information
Set Membership: |
|

RPS11
Genomic Map