Assay Details
| Gene Symbol: DHFR-TS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251715.1 | NP_001238644.1 | 2 - 3 | 581 | 93 | |
| GenBank mRNA | BT093772.1 | - | 2 - 3 | 599 | 93 | |
| S78087.1 | - | 2 - 3 | 581 | 93 | ||
More Information
Set Membership: |
|

DHFR-TS
Genomic Map