Assay Details
| Gene Symbol: DHFR-TS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251715.1 | NP_001238644.1 | 6 - 7 | 1160 | 72 | |
| GenBank mRNA | BT093771.1 | - | 4 - 5 | 307 | 72 | |
| S78087.1 | - | 6 - 7 | 1160 | 72 | ||
More Information
Set Membership: |
|

DHFR-TS
Genomic Map