Assay Details
| Gene Symbol: HEMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251808.1 | NP_001238737.1 | 1 - 2 | 263 | 71 | |
| GenBank mRNA | AK243972.1 | - | 1 - 2 | 272 | 71 | |
| AK285742.1 | - | 1 - 2 | 329 | 71 | ||
| U04525.1 | - | 1 - 2 | 263 | 71 | ||
More Information
Set Membership: |
|

HEMB
Genomic Map