Assay Details
| Gene Symbol: HEMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251808.1 | NP_001238737.1 | 3 - 4 | 455 | 73 | |
| GenBank mRNA | AK243972.1 | - | 3 - 4 | 464 | 73 | |
| AK285742.1 | - | 3 - 4 | 521 | 73 | ||
| U04525.1 | - | 3 - 4 | 455 | 73 | ||
More Information
Set Membership: |
|

HEMB
Genomic Map