Assay Details
Gene Symbol: AAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251355.1 | NP_001238284.1 | 4 - 5 | 355 | 72 | |
GenBank mRNA | AK246054.1 | - | 4 - 5 | 310 | 72 | |
U12735.1 | - | 4 - 5 | 355 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
