Assay Details
Gene Symbol: PIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248275.1 | NP_001235204.1 | 4 - 5 | 836 | 83 | |
GenBank mRNA | U27347.1 | - | 4 - 5 | 836 | 83 |
Gene Symbol: LOC100782155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253110.1 | NP_001240039.1 | 3 - 4 | 820 | 83 | |
GenBank mRNA | AK244476.1 | - | 3 - 4 | 846 | 83 | |
BT097900.1 | - | 1 - 2 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
