Assay Details
Gene Symbol: LOC547975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249912.1 | NP_001236841.1 | 3 - 4 | 203 | 97 | |
GenBank mRNA | AK244247.1 | - | 3 - 4 | 257 | 97 | |
AK285973.1 | - | 3 - 4 | 97 | |||
BT091245.1 | - | 3 - 4 | 334 | 97 | ||
X58245.1 | - | 3 - 4 | 203 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
