Assay Details
Gene Symbol: LOC547975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249912.1 | NP_001236841.1 | 6 - 7 | 382 | 73 | |
GenBank mRNA | AK244247.1 | - | 6 - 7 | 436 | 73 | |
AK285973.1 | - | 6 - 7 | 436 | 73 | ||
BT091245.1 | - | 6 - 7 | 513 | 73 | ||
X58245.1 | - | 6 - 7 | 382 | 73 |
Gene Symbol: LOC100500173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251010.1 | NP_001237939.1 | 2 - 3 | 73 | ||
GenBank mRNA | BT091038.1 | - | 6 - 7 | 445 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
