Assay Details
Gene Symbol: LOC547975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249912.1 | NP_001236841.1 | 7 - 8 | 473 | 96 | |
GenBank mRNA | AK244247.1 | - | 7 - 8 | 527 | 96 | |
AK285973.1 | - | 7 - 8 | 527 | 96 | ||
BT091245.1 | - | 7 - 8 | 604 | 96 | ||
X58245.1 | - | 7 - 8 | 473 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
