Assay Details
Gene Symbol: BMY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249318.1 | NP_001236247.1 | 2 - 3 | 565 | 79 | |
GenBank mRNA | AB004271.1 | - | 2 - 3 | 562 | 79 | |
AB189842.1 | - | 2 - 3 | 459 | 79 | ||
AB189843.1 | - | 2 - 3 | 462 | 79 | ||
AB189915.1 | - | 2 - 3 | 79 | |||
AB189916.1 | - | 2 - 3 | 459 | 79 | ||
AK243900.1 | - | 2 - 3 | 566 | 79 | ||
AK285807.1 | - | 2 - 3 | 566 | 79 | ||
D50866.1 | - | 2 - 3 | 530 | 79 | ||
DQ114423.1 | - | 2 - 3 | 456 | 79 | ||
M92090.1 | - | 2 - 3 | 557 | 79 | ||
X71419.1 | - | 2 - 3 | 565 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
