Assay Details
Gene Symbol: BMY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249318.1 | NP_001236247.1 | 5 - 6 | 1205 | 80 | |
GenBank mRNA | AB004271.1 | - | 5 - 6 | 1202 | 80 | |
AB189842.1 | - | 5 - 6 | 1099 | 80 | ||
AB189843.1 | - | 5 - 6 | 1102 | 80 | ||
AB189915.1 | - | 5 - 6 | 1099 | 80 | ||
AK243900.1 | - | 5 - 6 | 1206 | 80 | ||
AK285807.1 | - | 5 - 6 | 1206 | 80 | ||
D50866.1 | - | 5 - 6 | 1170 | 80 | ||
DQ114422.1 | - | 5 - 6 | 1096 | 80 | ||
DQ114423.1 | - | 5 - 6 | 1096 | 80 | ||
M92090.1 | - | 5 - 6 | 1197 | 80 | ||
X71419.1 | - | 5 - 6 | 1205 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
