Assay Details
| Gene Symbol: GPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249944.1 | NP_001236873.1 | 4 - 5 | 438 | 88 | |
| GenBank mRNA | X95582.1 | - | 4 - 5 | 438 | 88 | |
More Information
Set Membership: |
|

GPA2
Genomic Map