Assay Details
| Gene Symbol: SFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251399.1 | NP_001238328.1 | 4 - 5 | 806 | 101 | |
| GenBank mRNA | AM238658.1 | - | 4 - 5 | 806 | 101 | |
More Information
Set Membership: |
|

SFR2
Genomic Map