Assay Details
Gene Symbol: MYB55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT098318.1 | - | 5 - 6 | 713 | 113 | |
DQ822885.1 | - | 5 - 6 | 674 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
