Assay Details
Gene Symbol: BMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249421.1 | NP_001236350.1 | 2 - 3 | 731 | 85 | |
GenBank mRNA | AJ871579.1 | - | 2 - 3 | 731 | 85 | |
AK245180.1 | - | 1 - 2 | 159 | 85 | ||
AK245673.1 | - | 2 - 3 | 744 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
