Assay Details
| Gene Symbol: GDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249475.1 | NP_001236404.1 | 2 - 3 | 269 | 100 | |
| GenBank mRNA | AJ879895.1 | - | 2 - 3 | 269 | 100 | |
More Information
Set Membership: |
|

GDH1
Genomic Map