Assay Details
Gene Symbol: GDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249475.1 | NP_001236404.1 | 5 - 6 | 674 | 76 | |
GenBank mRNA | AJ879895.1 | - | 5 - 6 | 674 | 76 |
Gene Symbol: LOC100789509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252716.2 | NP_001239645.1 | 4 - 5 | 634 | 76 | |
GenBank mRNA | AK245891.1 | - | 4 - 5 | 699 | 76 | |
BT095808.1 | - | 4 - 5 | 636 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
