Assay Details
| Gene Symbol: GDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249475.1 | NP_001236404.1 | 6 - 7 | 794 | 88 | |
| GenBank mRNA | AJ879895.1 | - | 6 - 7 | 794 | 88 | |
| Gene Symbol: LOC100789509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252716.2 | NP_001239645.1 | 5 - 6 | 754 | 88 | |
| GenBank mRNA | AK245891.1 | - | 5 - 6 | 819 | 88 | |
| BT095808.1 | - | 5 - 6 | 756 | 88 | ||
More Information
Set Membership: |
|

GDH1
Genomic Map