Assay Details
Gene Symbol: GDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249475.1 | NP_001236404.1 | 8 - 9 | 1125 | 69 | |
GenBank mRNA | AJ879895.1 | - | 8 - 9 | 1125 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
