Assay Details
| Gene Symbol: LOC732542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249779.1 | NP_001236708.1 | 5 - 6 | 752 | 101 | |
| GenBank mRNA | AY954628.1 | - | 5 - 6 | 731 | 101 | |
| AY954629.1 | - | 5 - 6 | 752 | 101 | ||
More Information
Set Membership: |
|

LOC732542
Genomic Map