Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251443.1 | NP_001238372.1 | 3 - 4 | 560 | 66 | |
| GenBank mRNA | AY956421.1 | - | 3 - 4 | 560 | 66 | |
| Gene Symbol: VTE2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254567.1 | NP_001241496.1 | 2 - 3 | 340 | 66 | |
| GenBank mRNA | AK245240.1 | - | 2 - 3 | 385 | 66 | |
| DQ231059.1 | - | 2 - 3 | 340 | 66 | ||
More Information
Set Membership: |
|

HPT1
Genomic Map