Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251443.1 | NP_001238372.1 | 8 - 9 | 995 | 73 | |
| GenBank mRNA | AY956421.1 | - | 8 - 9 | 995 | 73 | |
| Gene Symbol: VTE2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254567.1 | NP_001241496.1 | 7 - 8 | 775 | 73 | |
| GenBank mRNA | AK245240.1 | - | 7 - 8 | 820 | 73 | |
| DQ231059.1 | - | 7 - 8 | 775 | 73 | ||
More Information
Set Membership: |
|

HPT1
Genomic Map