Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251443.1 | NP_001238372.1 | 12 - 13 | 1362 | 91 | |
| GenBank mRNA | AK245015.1 | - | 12 - 13 | 1383 | 91 | |
| AY956421.1 | - | 12 - 13 | 1362 | 91 | ||
| Gene Symbol: VTE2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK245240.1 | - | 11 - 12 | 1187 | 91 | |
More Information
Set Membership: |
|

HPT1
Genomic Map