Assay Details
Gene Symbol: NAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249971.2 | NP_001236900.1 | 2 - 3 | 482 | 102 | |
GenBank mRNA | BT098651.1 | - | 2 - 3 | 564 | 102 | |
DQ028774.1 | - | 2 - 3 | 484 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
