Assay Details
Gene Symbol: MYB65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244720.1 | - | 3 - 4 | 308 | 69 | |
DQ822969.1 | - | 2 - 3 | 214 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
