Assay Details
| Gene Symbol: MYB65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244720.1 | - | 4 - 5 | 376 | 87 | |
| DQ822969.1 | - | 3 - 4 | 282 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB65| Gene Symbol: MYB65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244720.1 | - | 4 - 5 | 376 | 87 | |
| DQ822969.1 | - | 3 - 4 | 282 | 87 | ||
Set Membership: |
|