Assay Details
| Gene Symbol: MYB65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244720.1 | - | 5 - 6 | 647 | 70 | |
| DQ822969.1 | - | 5 - 6 | 476 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB65| Gene Symbol: MYB65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244720.1 | - | 5 - 6 | 647 | 70 | |
| DQ822969.1 | - | 5 - 6 | 476 | 70 | ||
Set Membership: |
|