Assay Details
| Gene Symbol: LOC100170694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250891.1 | NP_001237820.1 | 1 - 2 | 335 | 63 | |
| GenBank mRNA | AK244916.1 | - | 1 - 2 | 340 | 63 | |
| AK245366.1 | - | 340 | 63 | |||
| EF440354.1 | - | 1 - 2 | 335 | 63 | ||
More Information
Set Membership: |
|

LOC100170694
Genomic Map