Assay Details
Gene Symbol: EN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248396.1 | NP_001235325.1 | 2 - 3 | 320 | 84 | |
GenBank mRNA | AK246074.1 | - | 2 - 3 | 361 | 84 | |
BT097673.1 | - | 2 - 3 | 358 | 84 | ||
EF521872.1 | - | 2 - 3 | 320 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
