Assay Details
Gene Symbol: EN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248396.1 | NP_001235325.1 | 4 - 5 | 489 | 72 | |
GenBank mRNA | AK246074.1 | - | 4 - 5 | 530 | 72 | |
BT097673.1 | - | 4 - 5 | 527 | 72 | ||
EF521872.1 | - | 4 - 5 | 489 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
