Assay Details
| Gene Symbol: EN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248396.1 | NP_001235325.1 | 8 - 9 | 804 | 90 | |
| GenBank mRNA | BT097674.1 | - | 3 - 4 | 173 | 90 | |
| EF521872.1 | - | 8 - 9 | 804 | 90 | ||
More Information
Set Membership: |
|

EN1
Genomic Map