Assay Details
| Gene Symbol: LOC100170729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251464.1 | NP_001238393.1 | 2 - 3 | 112 | 109 | |
| GenBank mRNA | AB333788.1 | - | 2 - 3 | 112 | 109 | |
More Information
Set Membership: |
|

LOC100170729
Genomic Map