Assay Details
Gene Symbol: LCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248258.1 | NP_001235187.1 | 4 - 5 | 481 | 87 | |
GenBank mRNA | AK243955.1 | - | 6 - 7 | 990 | 87 | |
AK244106.1 | - | 6 - 7 | 970 | 87 | ||
AK286157.1 | - | 7 - 8 | 1133 | 87 | ||
EU076433.1 | - | 4 - 5 | 481 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
