Assay Details
Gene Symbol: PHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251781.2 | NP_001238710.1 | 4 - 5 | 1068 | 61 | |
GenBank mRNA | AB359233.1 | - | 4 - 5 | 1119 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
