Assay Details
Gene Symbol: LOC100170691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250838.1 | NP_001237767.1 | 1 | 353 | 69 | |
GenBank mRNA | EU444602.1 | - | 1 | 353 | 69 |
Gene Symbol: LOC100305522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249610.1 | NP_001236539.1 | 1 | 362 | 69 | |
GenBank mRNA | BT089160.1 | - | 1 | 362 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
