Assay Details
| Gene Symbol: DGAT1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250755.1 | NP_001237684.1 | 1 | 251 | 81 | |
| GenBank mRNA | AB257590.1 | - | 1 | 251 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGAT1b| Gene Symbol: DGAT1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250755.1 | NP_001237684.1 | 1 | 251 | 81 | |
| GenBank mRNA | AB257590.1 | - | 1 | 251 | 81 | |
Set Membership: |
|