Assay Details
Gene Symbol: AOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249503.1 | NP_001236432.1 | 1 | 47 | 97 | |
GenBank mRNA | DQ288260.1 | - | 1 | 47 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
