Assay Details
Gene Symbol: MYB134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ822937.1 | - | 2 | 1743 | 82 |
Gene Symbol: MYB114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244635.1 | - | 8 | 2776 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
