Assay Details
| Gene Symbol: LOC547835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251747.1 | NP_001238676.1 | 1 | 152 | 86 | |
| GenBank mRNA | AK286853.1 | - | 1 | 187 | 86 | |
| U04526.1 | - | 1 | 152 | 86 | ||
More Information
Set Membership: |
|

LOC547835
Genomic Map