Assay Details
Gene Symbol: LOC100500155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250712.2 | NP_001237641.2 | 1 - 2 | 315 | 69 | |
GenBank mRNA | BT091008.1 | - | 1 - 2 | 317 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
