Assay Details
| Gene Symbol: LOC100500155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250712.2 | NP_001237641.2 | 2 - 3 | 449 | 109 | |
| GenBank mRNA | BT091008.1 | - | 2 - 3 | 451 | 109 | |
More Information
Set Membership: |
|

LOC100500155
Genomic Map