Assay Details
Gene Symbol: LOC100526854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248118.2 | NP_001235047.1 | 2 - 3 | 345 | 100 | |
GenBank mRNA | BT091654.1 | - | 2 - 3 | 347 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
