Assay Details
| Gene Symbol: LOC100804369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252785.1 | NP_001239714.1 | 5 - 6 | 509 | 91 | |
| GenBank mRNA | BT097411.1 | - | 5 - 6 | 509 | 91 | |
More Information
Set Membership: |
|

LOC100804369
Genomic Map