Assay Details
Gene Symbol: LOC100807528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253263.2 | NP_001240192.1 | 7 - 8 | 939 | 116 | |
GenBank mRNA | BT094052.1 | - | 7 - 8 | 941 | 116 | |
FJ594401.1 | - | 7 - 8 | 894 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
