Assay Details
Gene Symbol: LOC100786803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253326.1 | NP_001240255.1 | 1 | 219 | 84 | |
GenBank mRNA | BT095894.1 | - | 1 | 219 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT095894.1, NM_001253326.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
